missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ 2XYT Broth (Granulated Media)

Used in the growth and cultivation of microorganisms in the lab and include E. coli, bacteriophage, and other bacterial strains

Brand:  Fisher BioReagents™ BP9743-5

 View more versions of this product

Product Code. 15430685

  • 605.00 € / 5kg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Granulated Media
  • Formulation per liter: Casein Peptone [91079-40-2] 16g, Yeast Extract [8013-01-2] 10g, Sodium Chloride [7647-14-5] 5g
  • Store at room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solid
Microbiology Media
Poly Pail
Dehydrated 2XYT
Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L
5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers