missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ 2XYT Broth (Granulated Media)

Used in the growth and cultivation of microorganisms in the lab and include E. coli, bacteriophage, and other bacterial strains

69.40 € - 605.00 €

Specifications

Form Solid
Media Type Dehydrated 2XYT
Product Type Microbiology Media
Formulation Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L
View More Specs

Products 3
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
15420685
View Documents
Fisher BioReagents™
BP9743-2
2 kg Poly Pail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15430675
View Documents
Fisher BioReagents™
BP9743-500
500 g Poly Bottle
69.40 €
500g
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
 
15430685
View Documents
Fisher BioReagents™
BP9743-5
5 kg Poly Pail
605.00 €
5kg
In Stock
to see stock.
Add to basket
 
Description

Description

  • Granulated Media
  • Formulation per liter: Casein Peptone [91079-40-2] 16g, Yeast Extract [8013-01-2] 10g, Sodium Chloride [7647-14-5] 5g
  • Store at room temperature

Specifications

Specifications

Solid
Dehydrated 2XYT
Microbiology Media
Casein Peptone 16g/L, Yeast extract 10g/L, Sodium Chloride 5g/L
Videos
SDS
Documents

Documents

Product Certifications