missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Quantity | 250 mg |
Formula Weight | 238.67 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction