missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

59.60 € - 520.00 €

Chemical Identifiers

CAS 892502-09-9
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
InChI Key JGTYTUGTENJXBY-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
PubChem CID 6485349
IUPAC Name 3-(pyrazol-1-ylmethyl)aniline
SMILES C1=CC(=CC(=C1)N)CN2C=CC=N2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10662475
View Documents Promotion Details
Thermo Scientific Maybridge
CC56114CB
250 mg
59.60 €
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10759595
View Documents Promotion Details
Thermo Scientific Maybridge
CC56114DA
1 g
161.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10471694
View Documents Promotion Details
Thermo Scientific Maybridge
CC56114DE
5 g
520.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

892502-09-9
173.219
3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
3-(pyrazol-1-ylmethyl)aniline
C10H11N3
JGTYTUGTENJXBY-UHFFFAOYSA-N
6485349
C1=CC(=CC(=C1)N)CN2C=CC=N2

Specifications

892502-09-9
C10H11N3
JGTYTUGTENJXBY-UHFFFAOYSA-N
3-(pyrazol-1-ylmethyl)aniline
6485349
97%
97%
3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
C1=CC(=CC(=C1)N)CN2C=CC=N2
173.219
173.22
3-(1H-Pyrazol-1-ylmethyl)aniline
Videos
SDS
Documents

Documents

Product Certifications