missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 202.28 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction