missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid, ≥97%, Thermo Scientific™

98.20 € - 253.00 €

Chemical Identifiers

CAS 859851-00-6
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD08271950
InChI Key MOPCEJWYTICWJM-UHFFFAOYSA-N
Synonym 3-2-furyl-1-methyl-1h-pyrazole-5-carboxylic acid, 5-furan-2-yl-2-methylpyrazole-3-carboxylic acid, 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl, 3-fur-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1-methylpyrazole-5-carboxylic acid
PubChem CID 7537643
SMILES CN1N=C(C=C1C(O)=O)C1=CC=CO1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10507854
View Documents
Thermo Scientific Maybridge
CC47001CB
250 mg Amber Glass Bottle
98.20 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10787475
View Documents
Thermo Scientific Maybridge
CC47001DA
1 g Amber Glass Bottle
253.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

859851-00-6
192.17
MOPCEJWYTICWJM-UHFFFAOYSA-N
7537643
C9H8N2O3
MFCD08271950
3-2-furyl-1-methyl-1h-pyrazole-5-carboxylic acid, 5-furan-2-yl-2-methylpyrazole-3-carboxylic acid, 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl, 3-fur-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1-methylpyrazole-5-carboxylic acid
CN1N=C(C=C1C(O)=O)C1=CC=CO1

Specifications

859851-00-6
100.0
C9H8N2O3
3-2-furyl-1-methyl-1h-pyrazole-5-carboxylic acid, 5-furan-2-yl-2-methylpyrazole-3-carboxylic acid, 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl, 3-fur-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1-methylpyrazole-5-carboxylic acid
CN1N=C(C=C1C(O)=O)C1=CC=CO1
192.17
192.17
3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid
96.5
97%
MFCD08271950
MOPCEJWYTICWJM-UHFFFAOYSA-N
5-(furan-2-yl)-2-methylpyrazole-3-carboxylic acid
7537643
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications