Learn More
3,3,3-Trifluoro-1-propanol, 97%, Thermo Scientific Chemicals
91.60 € - 268.00 €
Chemical Identifiers
CAS | 2240-88-2 |
---|---|
Molecular Formula | C3H5F3O |
Molecular Weight (g/mol) | 114.067 |
MDL Number | MFCD00190649 |
InChI Key | HDBGBTNNPRCVND-UHFFFAOYSA-N |
Synonym | 3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane |
PubChem CID | 137512 |
IUPAC Name | 3,3,3-trifluoropropan-1-ol |
SMILES | C(CO)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323816
|
Thermo Scientific Alfa Aesar
L16879.03 |
1 g |
91.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333816
|
Thermo Scientific Alfa Aesar
L16879.06 |
5 g |
268.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
Chemical Identifiers
2240-88-2 | |
114.067 | |
HDBGBTNNPRCVND-UHFFFAOYSA-N | |
137512 | |
C(CO)C(F)(F)F |
C3H5F3O | |
MFCD00190649 | |
3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane | |
3,3,3-trifluoropropan-1-ol |
Specifications
2240-88-2 | |
100°C to 101°C | |
C3H5F3O | |
MFCD00190649 | |
1734777 | |
Fully miscible in water. | |
C(CO)C(F)(F)F | |
114.067 | |
114.07 | |
3,3,3-Trifluoro-1-propanol |
1.294 | |
37°C (98°F) | |
1.32 | |
UN1987 | |
3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane | |
HDBGBTNNPRCVND-UHFFFAOYSA-N | |
3,3,3-trifluoropropan-1-ol | |
137512 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only