Learn More
3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals
87.44 € - 860.28 €
Chemical Identifiers
CAS | 75-97-8 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
MDL Number | MFCD00008846 |
InChI Key | PJGSXYOJTGTZAV-UHFFFAOYSA-N |
Synonym | pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone |
PubChem CID | 6416 |
IUPAC Name | 3,3-dimethylbutan-2-one |
SMILES | CC(=O)C(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10477520
|
Thermo Scientific Acros
131251000 |
100 mL | Glass Bottle |
87.44 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103090
|
Thermo Scientific Acros
131255000 |
500 mL | Glass bottle |
237.18 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974821
|
Thermo Scientific Acros
131250025 |
2.5 L | Glass Bottle |
860.28 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-97-8 | |
100.16 | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
6416 | |
CC(=O)C(C)(C)C |
C6H12O | |
MFCD00008846 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
3,3-dimethylbutan-2-one |
Specifications
75-97-8 | |
Colorless to Yellow | |
106.0°C | |
Authentic | |
C6H12O | |
1.3950 to 1.3980 | |
01, 694 | |
0.8 | |
Solubility in water: very soluble. Other solubilities: soluble in alcohol,ether and acetone | |
CC(=O)C(C)(C)C | |
100.16 | |
100.16 | |
Liquid |
-52.5°C | |
0.8000g/mL | |
5°C | |
95% min. (GC) | |
(CH3)3CCOCH3 | |
MFCD00008846 | |
15, 7551 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
3,3-dimethylbutan-2-one | |
6416 | |
96% | |
3, 3-Dimethyl-2-butanone, 96% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-920-4
missing translation for 'rtecsNumber' : EL7700000
missing translation for 'tsca' : TSCA
- 3,3-Dimethyl-2-butanone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only