missing translation for 'onlineSavingsMsg'
Learn More

3-[4-Chloro-2-(trifluoromethyl)phenyl]propionic acid, 95%, Thermo Scientific™

44.30 € - 148.00 €

Chemical Identifiers

CAS 916420-46-7
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.617
MDL Number MFCD09025363
InChI Key OWXLPJQRNXEXHZ-UHFFFAOYSA-N
Synonym 3-4-chloro-2-trifluoromethyl phenyl propionic acid, 3-4-chloro-2-trifluoromethyl phenyl propanoic acid
PubChem CID 46737495
IUPAC Name 3-[4-chloro-2-(trifluoromethyl)phenyl]propanoic acid
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)CCC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428095
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32443.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15418095
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32443.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

916420-46-7
252.617
OWXLPJQRNXEXHZ-UHFFFAOYSA-N
46737495
C1=CC(=C(C=C1Cl)C(F)(F)F)CCC(=O)O
C10H8ClF3O2
MFCD09025363
3-4-chloro-2-trifluoromethyl phenyl propionic acid, 3-4-chloro-2-trifluoromethyl phenyl propanoic acid
3-[4-chloro-2-(trifluoromethyl)phenyl]propanoic acid

Specifications

916420-46-7
C10H8ClF3O2
3-4-chloro-2-trifluoromethyl phenyl propionic acid, 3-4-chloro-2-trifluoromethyl phenyl propanoic acid
C1=CC(=C(C=C1Cl)C(F)(F)F)CCC(=O)O
252.617
252.62
3-[4-Chloro-2-(trifluoromethyl)phenyl]propionic acid
87°C to 90°C
MFCD09025363
OWXLPJQRNXEXHZ-UHFFFAOYSA-N
3-[4-chloro-2-(trifluoromethyl)phenyl]propanoic acid
46737495
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications