missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chloro-3-fluorophenyl)propionic acid, 96%, Thermo Scientific Chemicals

75.30 € - 242.00 €

Chemical Identifiers

CAS 881189-65-7
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.609
MDL Number MFCD04116058
InChI Key GDXMVKNNJGWZFN-UHFFFAOYSA-N
Synonym 3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
PubChem CID 4680108
IUPAC Name 3-(4-chloro-3-fluorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)F)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15447137
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H34205.03
1 g
75.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
15457137
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H34205.06
5 g
242.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

881189-65-7
202.609
GDXMVKNNJGWZFN-UHFFFAOYSA-N
4680108
C1=CC(=C(C=C1CCC(=O)O)F)Cl
C9H8ClFO2
MFCD04116058
3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
3-(4-chloro-3-fluorophenyl)propanoic acid

Specifications

881189-65-7
C9H8ClFO2
3-4-chloro-3-fluorophenyl propionic acid, 3-4-chloro-3-fluorophenyl propanoic acid, benzenepropanoic acid, 4-chloro-3-fluoro, 3-4-chloro-3-fluoro-phenyl-propionic acid, 4-chloro-3-fluorobenzenepropanoic acid, 3-4-chloro-3-fluorophenyl propionicacid
C1=CC(=C(C=C1CCC(=O)O)F)Cl
202.609
202.61
3-(4-Chloro-3-fluorophenyl)propionic acid
80°C to 84°C
MFCD04116058
GDXMVKNNJGWZFN-UHFFFAOYSA-N
3-(4-chloro-3-fluorophenyl)propanoic acid
4680108
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only