missing translation for 'onlineSavingsMsg'
Learn More
3,4-(Methylenedioxy)benzylideneacetone, 98%, Thermo Scientific Chemicals Product Code.: 11392646

3,4-(Methylenedioxy)benzylideneacetone, 98%, Thermo Scientific Chemicals

Product Code. 11392646
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11392646

Brand: Thermo Scientific Alfa Aesar L08675.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-37-0
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00016907
InChI Key XIYPXOFSURQTTJ-NSCUHMNNSA-N
Synonym piperonalacetone, heliotropyl acetone, acetone, piperonylidene, piperonylideneacetone, piperonylidene acetone, unii-t43byj64eg, 3-buten-2-one, 4-1,3-benzodioxol-5-yl, ccris 6270, 3,4-methylenedioxy benzalacetone, 3,4-methylenedioxy benzylideneacetoneShow MoreShow Less
PubChem CID 6040503
IUPAC Name (E)-4-(1,3-benzodioxol-5-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC2=C(C=C1)OCO2

Specifications

Melting Point 108°C to 110°C
Quantity 50 g
Beilstein 83983
Formula Weight 190.2
Percent Purity 98%
Chemical Name or Material 3,4-(Methylenedioxy)benzylideneacetone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10117686 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberL08675.09,L08675.18
Lot NumberGBFA013453 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL08675.09,L08675.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-(Methylenedioxy)benzylideneacetone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.