missing translation for 'onlineSavingsMsg'
Learn More
3,4-Methylenedioxyphenylboronic acid, 98%, Thermo Scientific Chemicals Product Code.: 10721975

3,4-Methylenedioxyphenylboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10721975
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10721975

Brand: Thermo Scientific Acros 359580050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94839-07-3
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.94
MDL Number MFCD01009695
InChI Key CMHPUBKZZPSUIQ-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenylboronic acid, benzo d 1,3 dioxol-5-ylboronic acid, 3,4-methylenedioxy phenylboronic acid, 3,4-methylenedioxyphenyl boronic acid, 3,4-methylenedioxy benzeneboronic acid, 1,3-benzodioxole-5-boronic acid, 3,4-methylenedioxybenzeneboronic acid, 2h-1,3-benzodioxol-5-yl boronic acid, 2h-1,3-benzodioxol-5-yl-boranediol, 2h-1,3-benzodioxol-5-ylboronic acidShow MoreShow Less
PubChem CID 2734371
IUPAC Name 1,3-benzodioxol-5-ylboronic acid
SMILES B(C1=CC2=C(C=C1)OCO2)(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 165.94
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 4-Methylenedioxyphenylboronic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0408787 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number359580050,359580010
compliance-icons
Product Identifier
  • 3,4-Methylenedioxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Methylenedioxyphenylboronic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.