Learn More
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine, 97%, Thermo Scientific™
20.90 € - 108.00 €
Chemical Identifiers
CAS | 845885-86-1 |
---|---|
Molecular Formula | C10H14BNO2 |
Molecular Weight (g/mol) | 191.04 |
MDL Number | MFCD03425979 |
InChI Key | HDQULWCYQQMIJJ-UHFFFAOYSA-N |
Synonym | 3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester |
PubChem CID | 2794717 |
IUPAC Name | 3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine |
SMILES | CC1(C)COB(OC1)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10107392
|
Thermo Scientific Maybridge
MO01247DA |
1 g |
20.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10623075
|
Thermo Scientific Maybridge
MO01247DE |
5 g |
59.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10545184
|
Thermo Scientific Maybridge
MO01247EA |
10 g |
108.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
845885-86-1 | |
191.04 | |
HDQULWCYQQMIJJ-UHFFFAOYSA-N | |
2794717 | |
CC1(C)COB(OC1)C1=CC=NC=C1 |
C10H14BNO2 | |
MFCD03425979 | |
3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester | |
3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine |
Specifications
845885-86-1 | |
97% | |
C10H14BNO2 | |
3-5,5-dimethyl-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid neopentylglycol ester, pyridine-3-boronic acid neopentylglycol ester, pyridine-3-boronic acid, neopentyl ester, pyridine-3-boronic acid neopentyl glycol ester, 2-3-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, 5,5-dimethyl-2-pyridin-3-yl-1,3,2-dioxaborinane, 2-4-pyridil-5,5-dimethyl-1,3,2-dioxaboronane, pyridine-3-boronic acid neopentyl ester, pyridine-3-boronic acid, neopentylglycol ester | |
CC1(C)COB(OC1)C1=CC=NC=C1 | |
191.04 | |
191.04 | |
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine |
97.0 | |
Amber glass bottle | |
MFCD03425979 | |
HDQULWCYQQMIJJ-UHFFFAOYSA-N | |
3-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)pyridine | |
2794717 | |
97% |
Safety and Handling
GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.