missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

89.04 € - 151.76 €

Chemical Identifiers

CAS 20857-47-0
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00128904
InChI Key RQEOYYWUVYZZLL-UHFFFAOYSA-N
PubChem CID 2778311
IUPAC Name 3,5-bis(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318756
View Documents
Thermo Scientific Alfa Aesar
L19447.MD
250 mg
89.04 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11308756
View Documents
Thermo Scientific Alfa Aesar
L19447.03
1 g
151.76 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20857-47-0
215.098
RQEOYYWUVYZZLL-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)pyridine
C7H3F6N
MFCD00128904
2778311
C1=C(C=NC=C1C(F)(F)F)C(F)(F)F

Specifications

20857-47-0
117°C to 118°C
C7H3F6N
1684576
RQEOYYWUVYZZLL-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)pyridine
2778311
97%
35°C to 38°C
>100°C (212°F)
MFCD00128904
3,5-bis trifluoromethyl pyridine, pyridine, 3,5-bis trifluoromethyl, pubchem7698, acmc-1bncl, 3,5-bis-trifluoromethylpyridine, pyridine,3,5-bis trifluoromethyl
C1=C(C=NC=C1C(F)(F)F)C(F)(F)F
215.098
215.1
3,5-Bis(trifluoromethyl)pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.