Learn More
3,5-Dihydroxytoluene monohydrate, 97%, Thermo Scientific Chemicals
27.74 € - 237.18 €
Chemical Identifiers
CAS | 6153-39-5 |
---|---|
Molecular Formula | C7H10O3 |
Molecular Weight (g/mol) | 142.154 |
MDL Number | MFCD00149092 |
InChI Key | NBKPNAMTHBIMLA-UHFFFAOYSA-N |
Synonym | 5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate |
PubChem CID | 3083941 |
IUPAC Name | 5-methylbenzene-1,3-diol;hydrate |
SMILES | CC1=CC(=CC(=C1)O)O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468653
|
Thermo Scientific Alfa Aesar
A10440.06 |
5 g |
27.74 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478653
|
Thermo Scientific Alfa Aesar
A10440.14 |
25 g |
86.93 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488653
|
Thermo Scientific Alfa Aesar
A10440.22 |
100 g |
237.18 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6153-39-5 | |
142.154 | |
NBKPNAMTHBIMLA-UHFFFAOYSA-N | |
3083941 | |
CC1=CC(=CC(=C1)O)O.O |
C7H10O3 | |
MFCD00149092 | |
5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate | |
5-methylbenzene-1,3-diol;hydrate |
Specifications
6153-39-5 | |
147°C (5 mmHg) | |
MFCD00149092 | |
Air Sensitive | |
5-methylbenzene-1,3-diol hydrate, orcinol monohydrate, 5-methylresorcinol monohydrate, 3,5-dihydroxytoluene monohydrate, orcinol hydrate, resorcinol, 5-methyl-, monohydrate, 1,3-benzenediol, 5-methyl-, monohydrate, 1,3-dihydroxy-5-methylbenzene monohydrate, acmc-209mtl, 5-methylresorcinol hydrate | |
CC1=CC(=CC(=C1)O)O.O | |
142.154 | |
142.16 (124.14 Anhydrous) | |
3,5-Dihydroxytoluene monohydrate |
56°C to 60°C | |
C7H10O3 | |
5103474 | |
14,6864 | |
NBKPNAMTHBIMLA-UHFFFAOYSA-N | |
5-methylbenzene-1,3-diol;hydrate | |
3083941 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 207-984-2
missing translation for 'rtecsNumber' : VH2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only