missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dimethoxytoluene, 98%, Thermo Scientific Chemicals Product Code.: 11300244

3,5-Dimethoxytoluene, 98%, Thermo Scientific Chemicals

Product Code. 11300244
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11300244

Brand: Thermo Scientific Alfa Aesar L00809.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4179-19-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00015435
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5iShow MoreShow Less
PubChem CID 77844
IUPAC Name 1,3-dimethoxy-5-methylbenzene
SMILES COC1=CC(OC)=CC(C)=C1

Specifications

Density 1.039
Boiling Point 244°C
Flash Point 101°C (213°F)
Odor Aromatic
Refractive Index 1.5225
Quantity 25 g
Beilstein 2043558
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxytoluene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10218491 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberL00809.06,L00809.03,L00809.14
Lot Number10162498 Certificate TypeCertificate of Analysis Date16/06/2023 Catalog NumberL00809.06,L00809,L00809.03,L00809.14
Lot Number10218491 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00809.06,L00809.03,L00809.14
Lot Number10231807 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00809.06,L00809.03,L00809.14
Lot Number10194195 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00809.06,L00809.03,L00809.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dimethoxytoluene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.