missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

253.26 € - 633.00 €

Chemical Identifiers

CAS 181180-42-7
Molecular Formula C6H7F3O3S2
Molecular Weight (g/mol) 248.24
InChI Key ZMDCEXLCDOPKGH-UHFFFAOYSA-N
Synonym 2,3-dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 2,3dihydro-4-trifluoromethylsulfonyloxy-6h-thiopyran, 3,6-dihydro-2h-thiopyran-4-yltrifluoromethanesulfonate, trifluoro-methanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, trifluoromethanesulfonic acid 3,6-dihydro-2h-thiopyran-4-yl ester, methanesulfonic acid, trifluoro-, 3,6-dihydro-2h-thiopyran-4-yl esterShow MoreShow Less
PubChem CID 22625751
IUPAC Name 3,6-dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate
SMILES C1CSCC=C1OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11975761
SDS View Documents
Thermo Scientific Acros
439750010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11985761
SDS View Documents
Thermo Scientific Acros
439750050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,6-Dihydro-2H-thiopyran-4-yl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.