Learn More
3-Aminopyridine, 99%, Thermo Scientific Chemicals
43.82 € - 412.05 €
Chemical Identifiers
CAS | 462-08-8 |
---|---|
Molecular Formula | C5H6N2 |
Molecular Weight (g/mol) | 94.12 |
MDL Number | MFCD00006400 |
InChI Key | CUYKNJBYIJFRCU-UHFFFAOYSA-N |
Synonym | 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridineShow More |
PubChem CID | 10009 |
IUPAC Name | pyridin-3-amine |
SMILES | NC1=CC=CN=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420733
|
Thermo Scientific Alfa Aesar
A13216.18 |
50 g |
43.82 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430733
|
Thermo Scientific Alfa Aesar
A13216.30 |
250 g |
137.69 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410733
|
Thermo Scientific Alfa Aesar
A13216.0B |
1000 g |
412.05 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.
Solubility
It is soluble in water as well as soluble in alcohol, benzene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
Chemical Identifiers
462-08-8 | |
94.12 | |
CUYKNJBYIJFRCU-UHFFFAOYSA-N | |
10009 | |
NC1=CC=CN=C1 |
C5H6N2 | |
MFCD00006400 | |
3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridineShow More | |
pyridin-3-amine |
Specifications
462-08-8 | |
10 | |
88°C (190°F) | |
C5H6N2 | |
UN2671 | |
Hygroscopic | |
3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine | |
CUYKNJBYIJFRCU-UHFFFAOYSA-N | |
pyridin-3-amine | |
10009 | |
99% |
58°C to 63°C | |
250°C to 252°C | |
Unpleasant | |
MFCD00006400 | |
105692 | |
14,473 | |
It is soluble in water as well as soluble in alcohol,benzene. | |
NC1=CC=CN=C1 | |
94.12 | |
94.12 | |
3-Aminopyridine |
Safety and Handling
GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H300-H311-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
missing translation for 'einecsNumber' : 207-322-2
missing translation for 'rtecsNumber' : US1650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.