missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-cyanopyridine, 97%, Thermo Scientific™
318.00 € - 876.00 €
Chemical Identifiers
CAS | 13958-98-0 |
---|---|
Molecular Formula | C6H3BrN2 |
Molecular Weight (g/mol) | 183.01 |
InChI Key | VPDIXJCBBUZNPO-UHFFFAOYSA-N |
PubChem CID | 11074051 |
IUPAC Name | 3-bromopyridine-4-carbonitrile |
SMILES | C1=CN=CC(=C1C#N)Br |
Description
Chemical Identifiers
13958-98-0 | |
183.01 | |
11074051 | |
C1=CN=CC(=C1C#N)Br |
C6H3BrN2 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
3-bromopyridine-4-carbonitrile |
Specifications
13958-98-0 | |
96% min. (GC) | |
3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo | |
C1=CN=CC(=C1C#N)Br | |
183.01 | |
183.01 | |
3-Bromo-4-cyanopyridine |
91°C to 94°C | |
C6H3BrN2 | |
VPDIXJCBBUZNPO-UHFFFAOYSA-N | |
3-bromopyridine-4-carbonitrile | |
11074051 | |
97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Bromo-4-cyanopyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title