missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methoxypyridine, 97%, Thermo Scientific™

21.90 € - 72.43 €

Chemical Identifiers

CAS 50720-12-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00234169
InChI Key FZWUIWQMJFAWJW-UHFFFAOYSA-N
Synonym 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
PubChem CID 817163
IUPAC Name 3-bromo-5-methoxypyridine
SMILES COC1=CC(=CN=C1)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10386724
SDS View Documents
Thermo Scientific Acros
397630010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10764945
SDS View Documents
Thermo Scientific Acros
397630050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

50720-12-2
188.02
FZWUIWQMJFAWJW-UHFFFAOYSA-N
817163
COC1=CC(=CN=C1)Br
C6H6BrNO
MFCD00234169
3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
3-bromo-5-methoxypyridine

Specifications

50720-12-2
100.0
White to Yellow
Authentic
C6H6BrNO
3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
COC1=CC(=CN=C1)Br
188.02
188.02
Low Melting Solid
96.0
31°C to 35°C
102°C
96% min. (GC)
MFCD00234169
FZWUIWQMJFAWJW-UHFFFAOYSA-N
3-bromo-5-methoxypyridine
817163
97%
3-Bromo-5-methoxypyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 3-Bromo-5-methoxypyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.