missing translation for 'onlineSavingsMsg'
Learn More
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™ Product Code.: 10342005

3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™

Product Code. 10342005
10 g
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy

Product Code. 10342005

Brand: Thermo Scientific Maybridge CC25410EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4926-47-0
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
InChI Key APYSHMNJHJRIDR-UHFFFAOYSA-N
PubChem CID 327951
IUPAC Name 3-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=C(N2C=C1)Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Synonym 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromoimidazo[1, 2-a]pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.