Learn More
3-Chloro-2-fluorophenol, 97%, Thermo Scientific™
115.00 € - 378.00 €
Chemical Identifiers
CAS | 2613-22-1 |
---|---|
Molecular Formula | C6H4ClFO |
Molecular Weight (g/mol) | 146.545 |
MDL Number | MFCD01631572 |
InChI Key | PCHPYNHSMSAJEU-UHFFFAOYSA-N |
Synonym | 3-chloro-2-fluoro-phenol, phenol, 3-chloro-2-fluoro, acmc-20agxt, pubchem2855, 3-chloro-2-fluorophe, 2-fluoro-3-chloro-phenol, intermediates-zcf02697, phenol,3-chloro-2-fluoro, ksc557c2h, 3-chloro-2-fluorophenol |
PubChem CID | 2773712 |
IUPAC Name | 3-chloro-2-fluorophenol |
SMILES | C1=CC(=C(C(=C1)Cl)F)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Chloro-2-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
2613-22-1 | |
146.545 | |
PCHPYNHSMSAJEU-UHFFFAOYSA-N | |
2773712 | |
C1=CC(=C(C(=C1)Cl)F)O |
C6H4ClFO | |
MFCD01631572 | |
3-chloro-2-fluoro-phenol, phenol, 3-chloro-2-fluoro, acmc-20agxt, pubchem2855, 3-chloro-2-fluorophe, 2-fluoro-3-chloro-phenol, intermediates-zcf02697, phenol,3-chloro-2-fluoro, ksc557c2h, 3-chloro-2-fluorophenol | |
3-chloro-2-fluorophenol |
Specifications
2613-22-1 | |
190°C | |
C6H4ClFO | |
UN3261 | |
Insoluble in water. | |
C1=CC(=C(C(=C1)Cl)F)O | |
146.545 | |
146.55 | |
3-Chloro-2-fluorophenol |
34°C to 38°C | |
Phenol-like | |
MFCD01631572 | |
3-chloro-2-fluoro-phenol, phenol, 3-chloro-2-fluoro, acmc-20agxt, pubchem2855, 3-chloro-2-fluorophe, 2-fluoro-3-chloro-phenol, intermediates-zcf02697, phenol,3-chloro-2-fluoro, ksc557c2h, 3-chloro-2-fluorophenol | |
PCHPYNHSMSAJEU-UHFFFAOYSA-N | |
3-chloro-2-fluorophenol | |
2773712 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures