missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 200.65 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Chloromethyl)-5-(3-thienyl)-1, 2, 4-oxadiazole |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction