missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorothiophene, 96%, Thermo Scientific™
47.53 € - 187.00 €
Chemical Identifiers
CAS | 17249-80-8 |
---|---|
Molecular Formula | C4H3ClS |
Molecular Weight (g/mol) | 118.59 |
MDL Number | MFCD00043887 |
InChI Key | QUBJDMPBDURTJT-UHFFFAOYSA-N |
Synonym | thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h |
PubChem CID | 87017 |
IUPAC Name | 3-chlorothiophene |
SMILES | C1=CSC=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10642522
|
Thermo Scientific Acros
258800050 |
5 g | Glass bottle | ||||||
10478612
|
Thermo Scientific Acros
258800250 |
25 g | Glass bottle |
187.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
17249-80-8 | |
118.59 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
87017 | |
C1=CSC=C1Cl |
C4H3ClS | |
MFCD00043887 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
3-chlorothiophene |
Spezifikation
17249-80-8 | |
100.0 | |
1.2760g/mL | |
37°C | |
95% min. (GC) | |
1.5520 to 1.5540 | |
1.276 | |
Solubility in water: <0.2g/L (20°C). | |
C1=CSC=C1Cl | |
118.59 | |
118.59 | |
Liquid |
97.5 | |
Colorless to Yellow | |
137.0°C to 139.0°C | |
Authentic | |
C4H3ClS | |
MFCD00043887 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
3-chlorothiophene | |
87017 | |
96% | |
3-Chlorothiophene, 98% |
Sicherheit und Handhabung
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Flammable liquid and vapor.
May damage the unborn child.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Danger
missing translation for 'tsca' : TSCA
Recommended Storage : Product darkens on storage
RUO – Research Use Only