Learn More
3-Cyano-1-propylboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
80.10 € - 880.00 €
Chemical Identifiers
CAS | 238088-16-9 |
---|---|
Molecular Formula | C10H18BNO2 |
Molecular Weight (g/mol) | 195.07 |
MDL Number | MFCD09953480 |
InChI Key | LVMSRWXPZLRTIC-UHFFFAOYSA-N |
Synonym | 3-cyano-1-propylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 4-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 1,3,2-dioxaborolane-2-butanenitrile, 4,4,5,5-tetramethyl, 3-cyanoprop-1-ylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butyronitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl butanenitrile |
PubChem CID | 11183176 |
IUPAC Name | 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile |
SMILES | CC1(C)OB(CCCC#N)OC1(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11416230
|
Thermo Scientific Alfa Aesar
H27126.03 |
1 g |
80.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11426230
|
Thermo Scientific Alfa Aesar
H27126.06 |
5 g |
259.29 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436230
|
Thermo Scientific Alfa Aesar
H27126.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
238088-16-9 | |
195.07 | |
LVMSRWXPZLRTIC-UHFFFAOYSA-N | |
11183176 | |
CC1(C)OB(CCCC#N)OC1(C)C |
C10H18BNO2 | |
MFCD09953480 | |
3-cyano-1-propylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 4-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 1,3,2-dioxaborolane-2-butanenitrile, 4,4,5,5-tetramethyl, 3-cyanoprop-1-ylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butyronitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl butanenitrile | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile |
Specifications
238088-16-9 | |
MFCD09953480 | |
3-cyano-1-propylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 4-tetramethyl-1,3,2-dioxaborolan-2-yl butanenitrile, 1,3,2-dioxaborolane-2-butanenitrile, 4,4,5,5-tetramethyl, 3-cyanoprop-1-ylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl butyronitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl butanenitrile | |
CC1(C)OB(CCCC#N)OC1(C)C | |
195.07 | |
195.07 | |
3-Cyano-1-propylboronic acid pinacol ester |
C10H18BNO2 | |
UN3276 | |
LVMSRWXPZLRTIC-UHFFFAOYSA-N | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile | |
11183176 | |
96% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.