missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries 3-D Orbital Shaker Attachment

The 3-D Orbital Shaker Attachment for the Roto-Shake Genie or for the Enviro-Genie provides gentle yet thorough mixing of dishes, flasks, beakers and open containers.

Brand:  Scientific Industries SI-1150

 View more versions of this product

Product Code. 10073682

  • 358.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 3-D Orbital Shaker Attachment provides gentle yet thorough mixing of dishes, flasks, beakers and open containers
  • Moves at a 5° angle through a 360° rotation, providing a combination of vertical and horizontal motions
  • Ideal for applications such as gel and blot staining/blocking, sample washing, polymer resin preparation, and more
  • For use with the Roto-Shake Genie or the Enviro-Genie
Specifications

Specifications

1 kg
360°
3-D Orbital Shaker Attachment
1.4 kg
Gel and blot staining/blocking, sample washing, polymer resin preparation, and more
3-D Orbital Attachment, Non-skid Mat
Dishes, flasks, beakers, and open containers
3 lb.
Black
Roto-Shake Genie
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.