missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-5-methoxybenzeneboronic acid, 98%, Thermo Scientific™
92.20 € - 362.00 €
Chemical Identifiers
CAS | 609807-25-2 |
---|---|
Molecular Formula | C7H8BFO3 |
Molecular Weight (g/mol) | 169.946 |
MDL Number | MFCD07363780 |
InChI Key | MQFKLCWETPKBCH-UHFFFAOYSA-N |
Synonym | 3-fluoro-5-methoxybenzeneboronic acid, 3-fluoro-5-methoxyphenyl boronic acid, boronic acid, 3-fluoro-5-methoxyphenyl, 3-fluoro-5-methoxyphenylboronicacid, 3-fluoro-5-methoxy-phenyl boronic acid, acmc-1b9ik, 3-fluoro-5-methoxybenzeneboronicacid, 3-fluoro-5-methoxy phenylboronic acid, 3-fluoro-5-methoxyphenylboronic acid |
PubChem CID | 2763243 |
IUPAC Name | (3-fluoro-5-methoxyphenyl)boronic acid |
SMILES | B(C1=CC(=CC(=C1)F)OC)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
609807-25-2 | |
169.946 | |
MQFKLCWETPKBCH-UHFFFAOYSA-N | |
2763243 | |
B(C1=CC(=CC(=C1)F)OC)(O)O |
C7H8BFO3 | |
MFCD07363780 | |
3-fluoro-5-methoxybenzeneboronic acid, 3-fluoro-5-methoxyphenyl boronic acid, boronic acid, 3-fluoro-5-methoxyphenyl, 3-fluoro-5-methoxyphenylboronicacid, 3-fluoro-5-methoxy-phenyl boronic acid, acmc-1b9ik, 3-fluoro-5-methoxybenzeneboronicacid, 3-fluoro-5-methoxy phenylboronic acid, 3-fluoro-5-methoxyphenylboronic acid | |
(3-fluoro-5-methoxyphenyl)boronic acid |
Specifications
609807-25-2 | |
C7H8BFO3 | |
3-fluoro-5-methoxybenzeneboronic acid, 3-fluoro-5-methoxyphenyl boronic acid, boronic acid, 3-fluoro-5-methoxyphenyl, 3-fluoro-5-methoxyphenylboronicacid, 3-fluoro-5-methoxy-phenyl boronic acid, acmc-1b9ik, 3-fluoro-5-methoxybenzeneboronicacid, 3-fluoro-5-methoxy phenylboronic acid, 3-fluoro-5-methoxyphenylboronic acid | |
B(C1=CC(=CC(=C1)F)OC)(O)O | |
169.946 | |
169.95 | |
3-Fluoro-5-methoxybenzeneboronic acid |
168°C to 172°C | |
MFCD07363780 | |
MQFKLCWETPKBCH-UHFFFAOYSA-N | |
(3-fluoro-5-methoxyphenyl)boronic acid | |
2763243 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures