Learn More
3-Hexyn-2-one, 97%, Thermo Scientific Chemicals
85.20 € - 291.00 €
Chemical Identifiers
CAS | 1679-36-3 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.129 |
MDL Number | MFCD00041627 |
InChI Key | LTAPKZGQTMVYMX-UHFFFAOYSA-N |
Synonym | 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone |
PubChem CID | 137151 |
IUPAC Name | hex-3-yn-2-one |
SMILES | CCC#CC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307395
|
Thermo Scientific Alfa Aesar
L05315.04 |
2 g |
85.20 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317395
|
Thermo Scientific Alfa Aesar
L05315.09 |
10 g |
291.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1679-36-3 | |
96.129 | |
LTAPKZGQTMVYMX-UHFFFAOYSA-N | |
137151 | |
CCC#CC(=O)C |
C6H8O | |
MFCD00041627 | |
3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone | |
hex-3-yn-2-one |
Specifications
1679-36-3 | |
67°C to 68°C (50 mmHg) | |
C6H8O | |
MFCD00041627 | |
1699613 | |
LTAPKZGQTMVYMX-UHFFFAOYSA-N | |
hex-3-yn-2-one | |
137151 | |
97% |
0.88 | |
39°C (102°F) | |
1.441 | |
UN1992 | |
3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone | |
CCC#CC(=O)C | |
96.129 | |
96.13 | |
3-Hexyn-2-one |
Safety and Handling
GHS H Statement
H331-H318-H226-H302-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H300-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.