missing translation for 'onlineSavingsMsg'
Learn More
3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 11475570

3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11475570
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11475570

Brand: Thermo Scientific Alfa Aesar H25842.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.

Solubility
Soluble in water, DMSO, methanol.

Notes
Hygroscopic. Store under inert gas. Store away from water/ moisture, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18621-18-6
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD02683887
InChI Key UQUPQEUNHVVNKW-UHFFFAOYSA-N
Synonym 3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochlorideShow MoreShow Less
PubChem CID 2759290
IUPAC Name azetidin-3-ol;hydrochloride
SMILES C1C(CN1)O.Cl

Specifications

Melting Point 85°C to 90°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,DMSO,methanol.
Formula Weight 109.56
Percent Purity 97%
Chemical Name or Material 3-Hydroxyazetidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10211174 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25842.03,H25842.MD,H25842.06
Lot Number10168018 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25842.03,H25842.MD,H25842.06
Lot NumberY23C044 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25842.03,H25842.MD,H25842.06
Lot NumberP30B037 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25842.03,H25842.MD,H25842.06
Lot Number10137141 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25842.03,H25842.MD,H25842.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.