Learn More
3-Hydroxybenzoic acid, 99%, Thermo Scientific Chemicals
37.69 € - 321.60 €
Chemical Identifiers
CAS | 99-06-9 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002506 |
InChI Key | IJFXRHURBJZNAO-UHFFFAOYSA-N |
Synonym | m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova |
PubChem CID | 7420 |
ChEBI | CHEBI:30764 |
IUPAC Name | 3-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=CC(O)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10774051
|
Thermo Scientific Acros
120981000 |
100 g |
37.69 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10497880
|
Thermo Scientific Acros
120985000 |
500 g |
86.23 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10331334
|
Thermo Scientific Acros
120980025 |
2.5 kg |
321.60 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-06-9 | |
138.12 | |
IJFXRHURBJZNAO-UHFFFAOYSA-N | |
7420 | |
3-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002506 | |
m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova | |
CHEBI:30764 | |
OC(=O)C1=CC=CC(O)=C1 |
Specifications
99-06-9 | |
100.0 | |
White | |
Authentic | |
Plastic bottle | |
HOC6H4CO2H | |
10, 134 | |
Solubility in water: slightly soluble. Other solubilities: soluble in hot alcohol and ether | |
OC(=O)C1=CC=CC(O)=C1 | |
138.12 | |
CHEBI:30764 | |
99% | |
3-Hydroxybenzoic acid, 99% |
98.5 | |
201.0°C to 204.0°C | |
220°C | |
99% | |
C7H6O3 | |
MFCD00002506 | |
m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova | |
IJFXRHURBJZNAO-UHFFFAOYSA-N | |
3-hydroxybenzoic acid | |
7420 | |
138.12 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-726-5
missing translation for 'rtecsNumber' : DH1924980
missing translation for 'tsca' : TSCA
- 3-Hydroxybenzoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only