missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytyramine Hydrochloride, 99%, Thermo Scientific Chemicals

60.60 € - 375.87 €

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10152680
missing translation for 'viewDocuments'
Thermo Scientific Acros
122000100
10 g Glass bottle
60.60 €
10g
Please to purchase this item. Need a web account? Register with us today!
10428850
missing translation for 'viewDocuments'
Thermo Scientific Acros
122000250
25 g Glass bottle
112.56 €
25g
Please to purchase this item. Need a web account? Register with us today!
10454975
missing translation for 'viewDocuments'
Thermo Scientific Acros
122001000
100 g Glass bottle
375.87 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62-31-7
189.64
CTENFNNZBMHDDG-UHFFFAOYSA-N
65340
[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
C8H12ClNO2
MFCD00012898
dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride

Specifications

62-31-7
100.0
White
Authentic
C8H12ClNO2
MFCD00012898
15, 3470
Solubility in water: freely soluble. Other solubilities: soluble in methanol and hot 95% ethanol,practically insoluble in ether,chloroform,,benzene and toluene
[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
189.64
189.64
Crystalline Powder
98.5
241°C to 243°C
0.5% max. (105°C, 2 hrs)
99%
(HO)2C6H3CH2CH2NH2·HCl
13, I, 325
dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
CTENFNNZBMHDDG-UHFFFAOYSA-N
4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
65340
99%
3-Hydroxytyramine hydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-527-8

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.