missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

22.30 € - 234.00 €

Chemical Identifiers

CAS 1533748
MDL Number MFCD00004386
ChEBI CHEBI:64746
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469582
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10723.06
5 g
22.30 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11479582
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10723.14
25 g
72.10 €
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11489582
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10723.22
100 g
234.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1533748
MFCD00004386
CHEBI:64746

Specifications

1533748
C6H7K3O8
2209732
PJAHUDTUZRZBKM-UHFFFAOYSA-K
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
2735208
178.19
3-Methoxycinnamic acid, predominantly trans
117°C to 121°C
MFCD00004386
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
324.41
CHEBI:64746
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 228-049-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only