missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
3-Methylstyreneis used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methylstyreneis used as pharmaceutical intermediates.
Solubility
Soluble in Methanol, Ether, Benzene, Acetone, Ethanol, Heptane. Soluble in water (0.09 g/L) at 20°C.
Notes
Store at 4°C. Pr
3-Methylstyreneis used as pharmaceutical intermediates.
Solubility
Soluble in Methanol, Ether, Benzene, Acetone, Ethanol, Heptane. Soluble in water (0.09 g/L) at 20°C.
Notes
Store at 4°C. Pr
Specifications
Specifications
CAS | 100-80-1 |
Melting Point | -82°C to -81°C |
Density | 0.9 |
Boiling Point | 170°C to 171°C |
Flash Point | 51°C (123°F) |
Odor | Pungent |
Molecular Formula | C9H10 |
Refractive Index | 1.541 |
MDL Number | MFCD00008617 |
Quantity | 5 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'