Learn More
3-Picoline, 99%, Thermo Scientific Chemicals
26.23 € - 63.52 €
Chemical Identifiers
CAS | 108-99-6 |
---|---|
Molecular Formula | C6H7N |
Molecular Weight (g/mol) | 93.13 |
MDL Number | MFCD00006402 |
InChI Key | ITQTTZVARXURQS-UHFFFAOYSA-N |
Synonym | 3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine |
PubChem CID | 7970 |
ChEBI | CHEBI:39922 |
IUPAC Name | 3-methylpyridine |
SMILES | CC1=CN=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10051480
|
Thermo Scientific Acros
131132500 |
250 mL | Glass bottle |
26.23 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10347110
|
Thermo Scientific Acros
131135000 |
500 mL | Glass bottle |
38.39 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10387060
|
Thermo Scientific Acros
131130010 |
1 L | Glass bottle |
63.52 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246813
|
Thermo Scientific Acros
131130025 |
2.5 L | Glass bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-99-6 | |
93.13 | |
ITQTTZVARXURQS-UHFFFAOYSA-N | |
7970 | |
3-methylpyridine |
C6H7N | |
MFCD00006402 | |
3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine | |
CHEBI:39922 | |
CC1=CN=CC=C1 |
Specifications
108-99-6 | |
100.0 | |
Yellow | |
143.0°C to 144.0°C | |
Authentic | |
C6H7N | |
MFCD00006402 | |
15, 7513 | |
3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine | |
ITQTTZVARXURQS-UHFFFAOYSA-N | |
3-methylpyridine | |
7970 | |
93.13 | |
Liquid |
98.5 | |
-19.0°C | |
0.9500g/mL | |
38°C | |
98.5% min. (GC) | |
1.5050 to 1.5070 | |
20, 239 | |
0.95 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
CC1=CN=CC=C1 | |
93.13 | |
CHEBI:39922 | |
99% | |
3-Picoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-636-9
- 3-Picoline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 3-Methylpyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.