missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-(Trifluoromethoxy)acetophenone, 97%, Thermo Scientific Chemicals
46.10 € - 161.00 €
Chemical Identifiers
CAS | 170141-63-6 |
---|---|
Molecular Formula | C9H7F3O2 |
Molecular Weight (g/mol) | 204.15 |
MDL Number | MFCD00042403 |
InChI Key | UYHTUQHYGKAYJM-UHFFFAOYSA-N |
Synonym | 3'-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethanone, 3-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethan-1-one, 3-trifluoromethoxyacetophenone, 1-acetyl-3-trifluoromethoxy benzene, fxffor cv1, pubchem4297, intermediates-zcf02268 |
PubChem CID | 737161 |
IUPAC Name | 1-[3-(trifluoromethoxy)phenyl]ethanone |
SMILES | CC(=O)C1=CC=CC(OC(F)(F)F)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413387
|
Thermo Scientific Alfa Aesar
A17167.03 |
1 g | N/A | ||||||
11423387
|
Thermo Scientific Alfa Aesar
A17167.06 |
5 g |
161.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
170141-63-6 | |
204.15 | |
UYHTUQHYGKAYJM-UHFFFAOYSA-N | |
737161 | |
CC(=O)C1=CC=CC(OC(F)(F)F)=C1 |
C9H7F3O2 | |
MFCD00042403 | |
3'-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethanone, 3-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethan-1-one, 3-trifluoromethoxyacetophenone, 1-acetyl-3-trifluoromethoxy benzene, fxffor cv1, pubchem4297, intermediates-zcf02268 | |
1-[3-(trifluoromethoxy)phenyl]ethanone |
Specifications
170141-63-6 | |
49°C to 51°C (13 mmHg) | |
C9H7F3O2 | |
MFCD00042403 | |
3'-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethanone, 3-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethan-1-one, 3-trifluoromethoxyacetophenone, 1-acetyl-3-trifluoromethoxy benzene, fxffor cv1, pubchem4297, intermediates-zcf02268 | |
CC(=O)C1=CC=CC(OC(F)(F)F)=C1 | |
204.15 | |
204.15 | |
3'-(Trifluoromethoxy)acetophenone |
1.285 | |
83°C (181°F) | |
1.4515 | |
6196333 | |
UYHTUQHYGKAYJM-UHFFFAOYSA-N | |
1-[3-(trifluoromethoxy)phenyl]ethanone | |
737161 | |
97% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only