missing translation for 'onlineSavingsMsg'
Learn More

3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%, Thermo Scientific Chemicals

18.80 € - 213.00 €

Chemical Identifiers

CAS 190771-22-3
Molecular Formula C12H13NSi
Molecular Weight (g/mol) 199.328
MDL Number MFCD09260494
InChI Key QEZDMZVLNWSJSA-UHFFFAOYSA-N
Synonym 3-trimethylsilyl ethynyl benzonitrile, 3-2-trimethylsilyl ethynyl benzonitrile, benzonitrile,3-2-trimethylsilyl ethynyl, benzonitrile, 3-2-trimethylsilyl ethynyl, 3-trimethylsilanylethynylbenzonitrile, 3-trimethylsilanylethynyl-benzonitrile
PubChem CID 22570318
IUPAC Name 3-(2-trimethylsilylethynyl)benzonitrile
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15441128
View Documents
Thermo Scientific Alfa Aesar
H55012.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15421128
View Documents
Thermo Scientific Alfa Aesar
H55012.03
1 g
55.58 €
1g
Please to purchase this item. Need a web account? Register with us today!
15431128
View Documents
Thermo Scientific Alfa Aesar
H55012.06
5 g
213.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

190771-22-3
199.328
QEZDMZVLNWSJSA-UHFFFAOYSA-N
22570318
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
C12H13NSi
MFCD09260494
3-trimethylsilyl ethynyl benzonitrile, 3-2-trimethylsilyl ethynyl benzonitrile, benzonitrile,3-2-trimethylsilyl ethynyl, benzonitrile, 3-2-trimethylsilyl ethynyl, 3-trimethylsilanylethynylbenzonitrile, 3-trimethylsilanylethynyl-benzonitrile
3-(2-trimethylsilylethynyl)benzonitrile

Specifications

190771-22-3
C12H13NSi
3-trimethylsilyl ethynyl benzonitrile, 3-2-trimethylsilyl ethynyl benzonitrile, benzonitrile,3-2-trimethylsilyl ethynyl, benzonitrile, 3-2-trimethylsilyl ethynyl, 3-trimethylsilanylethynylbenzonitrile, 3-trimethylsilanylethynyl-benzonitrile
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
199.328
199.33
3-[(Trimethylsilyl)ethynyl]benzonitrile
42°C to 47°C
MFCD09260494
QEZDMZVLNWSJSA-UHFFFAOYSA-N
3-(2-trimethylsilylethynyl)benzonitrile
22570318
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.