Learn More
Nickel Electro™ Stainless Steel Laboratory Funnel
Use with or without filter membranes for gravity filtration as well as simple funnel procedures. Nickel Electro™ Stainless Steel Laboratory Funnel is vented to allow complete, uninhibited flow of liquids or solids.
19.85 € - 51.60 €
Product Code | Brand | Height (Metric) | Length (Metric) Stem | Diameter (Metric)Top | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Length (Metric) Stem | Diameter (Metric)Top | Price | Quantity & Availability | ||||
12963621
|
Nickel Electro™
FUNSS100H |
115 mm | 55 mm | 103 mm |
19.85 €
Each |
In Stock
Log in to see stock. ![]() |
||||
12983621
|
Nickel Electro™
FUNSS200H |
205 mm | 90 mm | 210 mm |
43.21 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||
12973621
|
Nickel Electro™
FUNSS150H |
158 mm | 75 mm | 165 mm |
51.60 €
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
- Durable stainless steel funnel, break-resistant, stain proof and easy to clean.
- May be used with or without filter membranes for gravity filtration as well as simple funnel procedures.
- Flute is vented to allow complete, uninhibited flow.
- Can be used with liquids or solids.
- The handle can also be used as a convenient hanging point when not in use.
- Smooth finish on internal surfaces making funnel easy to clean or sterilise.
Specifications
Stainless Steel | |
Handle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Laboratory Funnel