Learn More
Nickel Electro™ Stainless Steel Laboratory Funnel
Use with or without filter membranes for gravity filtration as well as simple funnel procedures. Nickel Electro™ Stainless Steel Laboratory Funnel is vented to allow complete, uninhibited flow of liquids or solids.
Brand:  Nickel Electro™ FUNSS150H
 View more versions of this product
Product Code. 12973621
  • 51.60 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Durable stainless steel funnel, break-resistant, stain proof and easy to clean.
  • May be used with or without filter membranes for gravity filtration as well as simple funnel procedures.
  • Flute is vented to allow complete, uninhibited flow.
  • Can be used with liquids or solids.
  • The handle can also be used as a convenient hanging point when not in use.
  • Smooth finish on internal surfaces making funnel easy to clean or sterilise.
Specifications
Specifications
165 mm
158 mm
75 mm
Stainless Steel
Handle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Laboratory Funnel > Diameter Outer: 150mm