Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ASTM Certified, Micrometer Pore Sizes
Brand:  RETSCH 60.159.000063
 View more versions of this product
Product Code. 10035051
  • 219.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
40mm
305 mm
Stainless Steel
63 μm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel