Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: 10 mm and Larger
251.00 € - 303.00 €
Specifications
Quantity | 1Each |
---|---|
Type | Test Sieve |
Certifications/Compliance | ISO 3310/1 |
Material | Stainless Steel |
For Use With (Equipment) | Shaker AS 300 |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10533342
|
RETSCH
60.158.106000 |
106 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10769892
|
RETSCH
60.158.100000 |
100 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10248931
|
RETSCH
60.158.010000 |
10 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10014951
|
RETSCH
60.158.011200 |
11.20 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10779892
|
RETSCH
60.158.112000 |
112 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10218691
|
RETSCH
60.158.125000 |
125 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10248731
|
RETSCH
60.158.012500 |
12.50 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10739892
|
RETSCH
60.158.013200 |
13.20 mm |
251.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10300912
|
RETSCH
60.158.018000 |
18 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10208881
|
RETSCH
60.158.019000 |
19 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10497571
|
RETSCH
60.158.014000 |
14 mm |
251.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10034791
|
RETSCH
60.158.020000 |
20 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10300862
|
RETSCH
60.158.022400 |
22.40 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10340762
|
RETSCH
60.158.016000 |
16 mm |
258.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10658571
|
RETSCH
60.158.025000 |
25 mm |
251.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10268971
|
RETSCH
60.158.028000 |
28 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10258971
|
RETSCH
60.158.026500 |
26.50 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10229021
|
RETSCH
60.158.031500 |
31.50 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10055071
|
RETSCH
60.158.035500 |
35.50 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10380282
|
RETSCH
60.158.037500 |
37.50 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10670082
|
RETSCH
60.158.040000 |
40 mm |
265.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
13514510
|
RETSCH
60.158.040000 |
4 mm |
303.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10406431
|
RETSCH
60.158.045000 |
45 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10467191
|
RETSCH
60.158.050000 |
50 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10779122
|
RETSCH
60.158.053000 |
53 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10789122
|
RETSCH
60.158.056000 |
56 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10555832
|
RETSCH
60.158.063000 |
63 mm |
265.00 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10034931
|
RETSCH
60.158.071000 |
71 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10044931
|
RETSCH
60.158.075000 |
75 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10258591
|
RETSCH
60.158.090000 |
90 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
10248591
|
RETSCH
60.158.080000 |
80 mm |
272.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||||
Specifications
1Each | |
ISO 3310/1 | |
Shaker AS 300 | |
40mm | |
305 mm | |
Stainless Steel |
Test Sieve | |
Stainless Steel | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |