Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
Brand:  RETSCH 60.138.014000
 View more versions of this product
Product Code. 10740125
  • 259.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
For separation, fractioning, particle size determination
Stainless Steel
14 mm
ISO 3310/2
40mm
305 mm
Stainless Steel