Learn More
Brannan™ ECO-therm Lab Thermometer
Green spirit thermometers filled with non toxic expansion liquid.
Brand:  Brannan™ 46/706/1
 View more versions of this product
Product Code. 15412695
  • 28.85 € / Pack of 10
Explore more special offers
Description
Description
  • Quality and resolution make these ideal substitute for general purpose mercury thermometers
  • Each thermometer is fitted with Brannan non roll end cap
  • Cap has integral ring to allow the thermometer to be suspended and is supplied in protective PVC packaging
Specifications
Specifications
Total Immersion
Glass
Laboratory Immersion Thermometer
-10°C
±1.5°C
305 mm
Green Spirit
150°C
-10°C to +150°C
1°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
Brannan™ ECO-therm Lab Thermometer > Temperature Range: -10°C to +150°C, Subdivisions: 1°, Total Immersion, Length: 305 mm