Learn More
3BDO, MedChemExpress
MedChemExpress 3BDO is a new mTOR activator which can also inhibit autophagy.
Brand: Medchem Express HY-U00434
This item is not returnable.
View return policy
Description
3BDO is a new mTOR activator which can also inhibit autophagy.Specifications
3BDO | |
Light Yellow | |
C18H17NO5 | |
DMSO : ≥ 100 mg/mL (305.50 mM) ∣Ethanol : 20 mg/mL (61.10 mM; Need ultrasonic) | |
327.33 | |
99.91% | |
Solid-Liquid Mixture |
890405-51-3 | |
Research | |
10 mg | |
O=C1OC(COC2=C([N+]([O-])=O)C=CC=C2)CC1CC3=CC=CC=C3 | |
327.33 | |
Research | |
Cancer-Kinase/protease |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3BDO, MedChemExpress