missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Lab Coat, Series 4440
Offers a barrier against non-hazardous particles and liquid splashes in light duty applications.
285.60 € - 423.00 €
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Unisex |
Material | Polypropylene |
Color | White |
Closure Type | Zipper |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15895111
|
3M™
7000089711 |
X-Large |
423.00 €
Pack of 50 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15759847
|
3M™
7000089709 |
Medium |
423.00 €
Pack of 50 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
15805121
|
3M™
7000089712 |
2X-Large |
423.00 €
Pack of 50 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
15769847
|
3M™
7000089714 |
4X-Large | N/A | N/A | N/A | |||||
12991335
|
3M™
7000089708 |
Small |
423.00 €
Pack of 50 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
15885111
|
3M™
7000089710 |
Large |
423.00 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
15815121
|
3M™
7000089713 |
3X-Large |
423.00 €
Pack of 50 |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
Description
- Breathable, lightweight material.
- Designed for light duty applications.
- Offers a barrier against non-hazardous particles and liquid splashes.
- Zip fastener.
- Anti-static treated.
- Knitted cuffs.
Specifications
General Purpose Lab Coat | |
Polypropylene | |
Zipper | |
Single-use | |
CE Category 1 | |
1.93 oz. |
Unisex | |
White | |
Knit | |
Serged | |
Catering, Cleanrooms, Electronics Manufacturing, General Maintenance, Laboratories | |
Microporous Laminate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Disposable Protective Lab Coat, Series 4440