Learn More
3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
Brand:  3M™ 7000001945
142.58 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15135157
  • 182.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fit Test Adapter
Quantitative Fit Testing
601
OSHA
Qualitative Fit Test Apparatus FT10 and FT30