Learn More
3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
13.33 € - 206.45 €
Specifications
Certifications/Compliance OSHA
View More Specs
Products 4
Product Code Brand For Use With (Application) Product Type Price Quantity & Availability  
Product Code Brand For Use With (Application) Product Type Price Quantity & Availability  
12993731
View Documents
3M™
7000051789
Provide secure hood environment Collar This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15749767
View Documents
3M™
7000051788
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Respirator Test Hood
13.33 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
12973731
View Documents
3M™
7000001897
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter Nebulizer This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15135157
View Documents
3M™
7000001945
Quantitative Fit Testing Fit Test Adapter
182.00 €
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
Description
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
Specifications
Specifications
OSHA