Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
152.00 € - 240.00 €
Specifications
Assigned Protection Factor (APF) 1,000
Product Type Outer Shroud
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
For Use With (Equipment) M-400 helmet
View More Specs
Products 2
Product Code Brand Style Price Quantity & Availability  
Product Code Brand Style Price Quantity & Availability  
15700078
View Documents
3M™
7100137850
Flame Resistant
240.00 €
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
15790068
View Documents
3M™
7100137905
High Durability
152.00 €
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
  • 3M Catalog# M-445, M-446, M-447, M-448
Specifications
Specifications
1,000
Outer Shroud
Cleaning, electrical, facility maintenance, painting, sanding, welding
M-400 helmet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Versaflo™ M-400 Series PAPR Outer Shroud