Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
Brand: 3M™ 7100137850
201.32 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
- 3M Catalog# M-445, M-446, M-447, M-448
Specifications
1,000 | |
Outer Shroud | |
M-400 helmet | |
Flame Resistant |
Nomex | |
Cleaning, electrical, facility maintenance, painting, sanding, welding | |
M-447 |
Videos
-
3M Powered and supplied air products
Product Content Correction
Product Title
3M™ Versaflo™ M-400 Series PAPR Outer Shroud > Style: Flame Resistant
Spot an opportunity for improvement?Share a Content Correction