missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

41.90 € - 620.00 €

Chemical Identifiers

CAS 160388-53-4
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD07186451
InChI Key ZHQQRHUITAFMTC-UHFFFAOYSA-N
PubChem CID 6484268
IUPAC Name 4-(pyrazol-1-ylmethyl)benzoic acid
SMILES C1=CN(N=C1)CC2=CC=C(C=C2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10634785
View Documents
Thermo Scientific Maybridge
CC56001CB
250 mg
41.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10285574
View Documents
Thermo Scientific Maybridge
CC56001DA
1 g
111.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10443204
View Documents
Thermo Scientific Maybridge
CC56001EA
10 g
620.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

160388-53-4
202.213
ZHQQRHUITAFMTC-UHFFFAOYSA-N
4-(pyrazol-1-ylmethyl)benzoic acid
C11H10N2O2
MFCD07186451
6484268
C1=CN(N=C1)CC2=CC=C(C=C2)C(=O)O

Specifications

160388-53-4
100.0
Amber Glass Bottle
MFCD07186451
ZHQQRHUITAFMTC-UHFFFAOYSA-N
4-(pyrazol-1-ylmethyl)benzoic acid
6484268
97%
96.5
97%
C11H10N2O2
4-1h-pyrazol-1-ylmethyl benzoic acid, 4-pyrazol-1-ylmethyl-benzoic acid, 4-pyrazol-1-ylmethyl benzoic acid, benzoicacid, 4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl benzoic acid, 4-1h-pyrazol-1-yl methyl benzoic acid, 4-1h-pyrazole-1-ylmethyl benzoic acid, benzoicacid,4-1h-pyrazol-1-ylmethyl, benzoic acid, 4-1h-pyrazol-1-ylmethyl
C1=CN(N=C1)CC2=CC=C(C=C2)C(=O)O
202.213
202.21
4-(1H-Pyrazol-1-ylmethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.