missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 4-(1H-pyrrol-1-yl)benzonitrile |
Formula Weight | 168.2 |
Percent Purity | 97% |
Chemical Name or Material | 4-(1H-pyrrol-1-yl)benzonitrile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction